Bis(tri-tert-butylphosphine)palladium(0) 98%

Supplier: Ambeed
Warning

Synonyms: Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)

A134602-10G A134602-1G A134602-5G
103802-028EA 41 USD
103802-028 76688-762 77314-792 77314-794
Bis(tri-tert-butylphosphine)palladium(0) 98%
Bis(tri-tert-butylphosphine)palladium(0)

Precaución: for Research Use Only

Formula: C₂₄H₅₄P₂Pd
MW: 511.05 g/mol
Melting Pt: 258…272 °C
Flash Pt: 94.6 °C
Storage Temperature: Freezer
MDL Number: MFCD03094580
CAS Number: 53199-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR