Bis(triphenylphosphine)nickel(II) chloride 98%

Supplier: Ambeed
Warning

Synonyms: Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride

A200982-100G A200982-10G A200982-1G A200982-500G A200982-25G A200982-5G A200982-1000G
103847-892EA 6 USD
103847-892 76999-318 76999-320 76999-322 76999-324 77085-956 77085-958 77600-640
Bis(triphenylphosphine)nickel(II) chloride 98%
Bis(triphenylphosphine)nickel(II) chloride

Precaución: for Research Use Only

Formula: C₃₆H₃₂Cl₂NiP₂
MW: 656.19 g/mol
Melting Pt: >300 °C
Flash Pt: 181.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009592
CAS Number: 14264-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR