Bis(tricyclohexylphosphine)nickel(II) dichloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bis(tricyclohexylphosphine)nickel(II) chloride, Dichlorobis(tricyclohexylphosphine)nickel(II)
A157054-100G
A157054-250MG
A157054-25G
A157054-1G
A157054-5G
103804-954EA
6
USD
103804-954
103843-156
77162-998
77163-000
77163-002
77318-968
77318-970
Bis(tricyclohexylphosphine)nickel(II) dichloride 97%
Bis(tricyclohexylphosphine)nickel(II) dichloride
Precaución: Inert atmosphere, room temperature
Formula:
C₃₆H₆₆Cl₂NiP₂ Boiling Pt: 383.4 °C (760 mm Hg) Melting Pt: 284 °C Flash Pt: 195.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD11973802 CAS Number: 19999-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...