Bis(2-pyridylmethyl)amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
alpha,alpha'-Iminodi(2-picoline), Bis(pyridin-2-ylmethyl)amine, 2,2'-Dipicolylamine
A108473-100G
A108473-10G
A108473-25G
A108473-500G
A108473-5G
A108473-1G
103797-354EA
5
USD
103797-354
103823-134
77245-568
77245-570
77245-572
77245-574
77283-748
77309-976
Bis(2-pyridylmethyl)amine 97%
Bis(2-pyridylmethyl)amine
Precaución: for Research Use Only
Formula:
C₁₂H₁₃N₃ Boiling Pt: 322 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00129044 CAS Number: 1539-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...