Bis(diphenylphosphino)methane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
[(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM
A118894-100G
A118894-10G
A118894-25G
A118894-500G
A118894-1G
A118894-5G
103799-126EA
5
USD
103799-126
76688-022
76688-024
76688-026
76688-028
77297-634
77311-800
Bis(diphenylphosphino)methane 98%
Bis(diphenylphosphino)methane
Precaución: for Research Use Only
Formula:
C₂₅H₂₂P₂ MW: 384.4 g/mol Boiling Pt: 497.2 °C (760 mm Hg) Melting Pt: 118…119 °C Flash Pt: 270.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003537 CAS Number: 2071-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...