Bis(dibenzylideneacetone)palladium 98%

Supplier: Ambeed
Warning

Synonyms: Bis(dibenzylideneacetone)palladium(0)

A107013-10G A107013-250MG A107013-25G A107013-5G
103797-114EA 23 USD
103797-114 103822-872 76994-970 76994-972 76994-974 77309-742
Bis(dibenzylideneacetone)palladium 98%
Bis(dibenzylideneacetone)palladium

Precaución: for Research Use Only

Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
Melting Pt: 150 °C
Flash Pt: 176.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR