Copper(II) acetylacetonate 97%

Supplier: Ambeed
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

A124434-1000G A124434-100G A124434-25G
76688-284EA 82 USD
76688-284 76688-286 76688-288 76688-290
Copper(II) acetylacetonate 97%
Copper(II) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄CuO₄
MW: 261.76 g/mol
Melting Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00000016
CAS Number: 13395-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR