Bis(acetonitrile)palladium(II) dichloride 98%

Supplier: Ambeed
Danger

Synonyms: Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)

A148299-10G A148299-250MG A148299-1G A148299-5G
103803-910EA 59 USD
103803-910 103842-026 76997-230 76997-232 77317-474 77317-476
Bis(acetonitrile)palladium(II) dichloride 98%
Bis(acetonitrile)palladium(II) dichloride

Precaución: for Research Use Only

Formula: C₄H₆Cl₂N₂Pd
MW: 259.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013122
CAS Number: 14592-56-4
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR