Bis(4-nitrophenyl)carbonate 97%

Supplier: Ambeed
Warning

A133221-100G A133221-10G A133221-500G A133221-25G A133221-5G
103808-456EA 320 USD
103808-456 77095-040 77095-042 77095-044 77314-484 77314-486
Bis(4-nitrophenyl)carbonate 97%
Bis(4-nitrophenyl)carbonate

Precaución: for Research Use Only

Formula: C₁₃H₈N₂O₇
Boiling Pt: 475.9 °C (760 mm Hg)
Melting Pt: 136…139 °C
Flash Pt: 217.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007322
CAS Number: 5070-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR