4,4'-Dimethoxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: Bis(4-methoxyphenyl)methanone

A275669-100G A275669-10G A275669-500G A275669-25G A275669-5G
103853-200EA 6 USD
103853-200 77032-036 77032-038 77032-040 77093-058 77305-210
4,4'-Dimethoxybenzophenone 98%
4,4'-Dimethoxybenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Melting Pt: 141…143 °C
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00008404
CAS Number: 90-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR