4,4'-Dimethoxydiphenylamine 98%

Supplier: Ambeed
Warning

Synonyms: Bis(4-methoxyphenyl)amine

A187910-25G A187910-5G A187910-100G A187910-1G A187910-10G
103816-050EA 7 USD
103816-050 103846-644 77084-360 77084-362 77287-432 77302-108 77615-590
4,4'-Dimethoxydiphenylamine 98%
4,4'-Dimethoxydiphenylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₂
MW: 229.28 g/mol
Boiling Pt: 371.1 °C (760 mm Hg)
Melting Pt: 101…102 °C
Flash Pt: 150.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00014895
CAS Number: 101-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR