Bis(4-methoxybenzyl)amine hydrochloride 97%

Supplier: Ambeed
Warning

A145739-100G A145739-10G A145739-250MG A145739-25G A145739-1G A145739-5G
103803-576EA 9 USD
103803-576 103841-658 77162-558 77162-560 77162-562 77299-472 77316-964 77316-966
Bis(4-methoxybenzyl)amine hydrochloride 97%
Bis(4-methoxybenzyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₀ClNO₂
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD07170733
CAS Number: 854391-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR