Bis(4-methoxybenzyl)amine 98%

Supplier: Ambeed
Danger

A594431-10G A594431-1G A594431-25G A594431-5G
103828-948EA 5 USD
103828-948 103840-088 76790-514 76844-234 76844-236 76844-238
Bis(4-methoxybenzyl)amine 98%
Bis(4-methoxybenzyl)amine

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₂
MW: 257.333 g/mol
Boiling Pt: 384 °C (760 mm Hg)
Melting Pt: 35…37 °C
Flash Pt: 161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00277836
CAS Number: 17061-62-0
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR