4,4'-Diaminobenzophenone 97%

Supplier: Ambeed
Warning

A122671-100G A122671-100MG A122671-250MG A122671-25G A122671-1G A122671-5G A122671-10G
103800-254EA 8 USD
103800-254 76810-596 76810-598 76810-600 76810-602 77312-384 77312-386 77566-324
4,4'-Diaminobenzophenone 97%
4,4'-Diaminobenzophenone

Precaución: For research use only.

Formula: C₁₃H₁₂N₂O
Boiling Pt: 452.8 °C (760 mm Hg)
Melting Pt: 243…247 °C
Flash Pt: 227.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00038138
CAS Number: 611-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR