3,3'-Dichlorobenzophenone 95%

Supplier: Ambeed
Warning

Synonyms: 3,3-DBP, Bis(3-chlorophenyl) ketone, Bis(3-chlorophenyl)methanone

A316158-250MG A316158-5G
103825-092EA 107 USD
103825-092 103836-018 77568-636 77571-894
3,3'-Dichlorobenzophenone 95%
3,3'-Dichlorobenzophenone

Precaución: For research use only. Sealed in dry, at room temperature.

Formula: C₁₃H₈Cl₂O
MW: 251.11 g/mol
Boiling Pt: 381.7 °C (760 mm Hg)
Flash Pt: 161.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00045192
CAS Number: 7094-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR