Bis(3,5-dimethylphenyl)phosphine oxide 96%
Supplier: Ambeed
|
Warning
|
A120862-10G
A120862-250MG
A120862-25G
A120862-1G
A120862-5G
A120862-100G
103800-002EA
13
USD
103800-002
77026-408
77026-410
77026-412
77312-108
77312-110
77871-533
Bis(3,5-dimethylphenyl)phosphine oxide 96%
Bis(3,5-dimethylphenyl)phosphine oxide
Precaución: for Research Use Only
Formula:
C₁₆H₁₉OP Boiling Pt: 414.8 °C (760 mm Hg) Flash Pt: 204.691 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458884 CAS Number: 187344-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...