Bis(2-chloroethyl)ammonium chloride 98%

Supplier: Ambeed
Danger

Synonyms: Bis(2-chloroethyl)amine hydrochloride

A104069-100G A104069-25G A104069-500G
103807-890EA 84 USD
103807-890 76869-294 76869-296 76869-298
Bis(2-chloroethyl)ammonium chloride 98%
Bis(2-chloroethyl)ammonium chloride

Precaución: for Research Use Only

Formula: C₄H₁₀Cl₃N
MW: 178.49 g/mol
Melting Pt: 212…214 °C
Flash Pt: 77.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00012515
CAS Number: 821-48-7
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR