Disuccinimidyl Succinate 97%

Supplier: Ambeed
Warning

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) succinate, Succinic acid Disuccinimidyl ester

A125401-250MG A125401-25G
103800-674EA 82 USD
103800-674 76797-636 77878-917
Disuccinimidyl Succinate 97%
Disuccinimidyl Succinate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₈
MW: 312.23 g/mol
Boiling Pt: 471.6 °C (760 mm Hg)
Flash Pt: 239 °C
Storage Temperature: Freezer
MDL Number: MFCD00228233
CAS Number: 30364-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR