DSS (Disuccinimidyl suberate) 97%

Supplier: Ambeed
Warning

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) octanedioate, Suberic acid bis(N-hydroxysuccinimide ester), Disuccinimidyl octanedioate, DSS

A211607-100MG A211607-10G A211607-250MG A211607-1G A211607-100G
103799-490EA 6 USD
103799-490 103818-122 103848-816 76800-652 76800-654 76800-656 77087-238 77626-072
DSS (Disuccinimidyl suberate) 97%
DSS (Disuccinimidyl suberate)

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₈
MW: 368.34 g/mol
Boiling Pt: 513.5 °C (760 mm Hg)
Melting Pt: 169…170 °C
Flash Pt: 264.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00049059
CAS Number: 68528-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR