Bis(2,4-dimethoxybenzyl)amine 97%

Supplier: Ambeed
Warning

A277335-10G A277335-250MG A277335-1G A277335-25G A277335-100G A277335-500G
103813-352EA 8 USD
103813-352 103853-286 76692-970 76692-972 77305-256 77305-258 77593-708 77626-432
Bis(2,4-dimethoxybenzyl)amine 97%
Bis(2,4-dimethoxybenzyl)amine

Precaución: for Research Use Only

Formula: C₁₈H₂₃NO₄
Boiling Pt: 443.5 °C (760 mm Hg)
Flash Pt: 188.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00040480
CAS Number: 20781-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR