Bis(2,4,6-trichlorophenyl) propanedioate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Bis(2,4,6-trichlorophenyl) malonate
A274698-10G
A274698-250MG
A274698-1G
A274698-25G
A274698-5G
103813-208EA
8
USD
103813-208
103853-136
76802-088
76802-090
77092-962
77092-964
77092-966
Bis(2,4,6-trichlorophenyl) propanedioate 98%
Bis(2,4,6-trichlorophenyl) propanedioate
Precaución: for Research Use Only
Formula:
C₁₅H₆Cl₆O₄ MW: 462.93 g/mol Boiling Pt: 559.6 °C (760 mm Hg) Melting Pt: 153…157 °C Flash Pt: 213 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00858973 CAS Number: 15781-70-1 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...