Bis(2,4,4-trimethylpentyl)phosphinic acid 90%
Supplier: Ambeed
|
Danger
|
A618704-5G
A618704-1G
A618704-25G
A618704-100G
A618704-500G
A618704-1000G
103840-200EA
7
USD
103840-200
76844-372
77307-954
77307-956
77551-856
77569-176
77573-012
Bis(2,4,4-trimethylpentyl)phosphinic acid 90%
Bis(2,4,4-trimethylpentyl)phosphinic acid
Precaución: For research use only.
Formula:
C₁₆H₃₅O₂P MW: 290.42 g/mol Boiling Pt: 269.9 °C (760 mm Hg) Flash Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD28145690 CAS Number: 83411-71-6 UN: 3082 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...