Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat

A108943-100MG A108943-250MG
103797-408EA 198 USD
103797-408 76970-578 77565-916
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate

Precaución: For research use only.

Formula: C₁₆H₂₄BF₄Rh
MW: 406.072 g/mol
Melting Pt: 210 °C
Storage Temperature: Freezer
MDL Number: MFCD00075045
CAS Number: 35138-22-8
UN: 2921
ADR: 8,4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR