Methoxy(cyclooctadiene)iridium(I) dimer 97%

Supplier: Ambeed
Warning

A190275-100MG A190275-250MG A190275-10G
103816-276EA 267 USD
103816-276 103846-878 76974-820 77550-214 77881-419
Methoxy(cyclooctadiene)iridium(I) dimer 97%
Methoxy(cyclooctadiene)iridium(I) dimer

Precaución: for Research Use Only

Formula: C₁₈H₃₀Ir₂O₂
Melting Pt: 154…177 °C
Storage Temperature: Freezer
MDL Number: MFCD08459360
CAS Number: 12148-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR