D-Biotin-N-succinimidyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide
A209315-100MG
A209315-1MG
A209315-250MG
A209315-1G
A209315-5G
103817-934EA
21
USD
103817-934
103848-620
76691-416
76691-418
76691-420
77086-960
77086-962
D-Biotin-N-succinimidyl ester 97%
D-Biotin-N-succinimidyl ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₉N₃O₅S Melting Pt: 212…214 °C Storage Temperature: Freezer |
MDL Number:
MFCD00078531 CAS Number: 35013-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...