Benzoyl-β-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Betamipron, 3-Benzamidopropanoic acid

A913610-100G A913610-100MG A913610-10G A913610-500G A913610-25G
103841-386EA 9 USD
103841-386 76698-678 76698-680 76698-682 76698-684 76845-662
Benzoyl-β-alanine 98%
Benzoyl-beta-alanine

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
Boiling Pt: 469.2 °C (760 mm Hg)
Melting Pt: 132 °C at N/A mmHg
Flash Pt: 237.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00037293
CAS Number: 3440-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR