Betahistine dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
A120842-100G
A120842-100MG
A120842-10G
A120842-25G
A120842-1G
A120842-5G
103799-998EA
24
USD
103799-998
77189-654
77189-656
77189-658
77189-660
77297-794
77297-796
Betahistine dihydrochloride 98%
Betahistine dihydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂·2HCl Boiling Pt: 210.9 °C (760 mm Hg) Melting Pt: 150…154 °C Flash Pt: 96.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012813 CAS Number: 5579-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...