Benzyltrimethylammonium dichloroiodate 98%
Supplier: Ambeed
|
Warning
|
A165770-100G
A165770-10G
A165770-500G
A165770-1G
A165770-5G
A165770-25G
103805-994EA
6
USD
103805-994
103844-248
76690-026
76690-028
76690-030
77300-796
77300-798
77320-470
Benzyltrimethylammonium dichloroiodate 98%
Benzyltrimethylammonium dichloroiodate
Precaución: for Research Use Only
Formula:
C₁₀H₁₆Cl₂IN MW: 348.06 g/mol Melting Pt: 124…126 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075259 CAS Number: 114971-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...