Benzyl methacrylate 98% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 50 ppm
A163971-100G
A163971-500G
A163971-25G
A163971-1000G
103808-936EA
70
USD
103808-936
76689-928
76689-930
77300-686
77320-186
Benzyl methacrylate 98% stabilized
Benzyl methacrylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂O₂ Boiling Pt: 247 °C (760 mm Hg) Flash Pt: 124.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00053684 CAS Number: 2495-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...