Benzyl carbazate 97%

Supplier: Ambeed
Warning

A168759-25G A168759-100G A168759-10G A168759-500G
103809-002EA 179 USD
103809-002 77081-634 77191-350 77191-352 77191-354
Benzyl carbazate 97%
Benzyl carbazate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
Melting Pt: 65…68 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041890
CAS Number: 5331-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR