Benzyl carbamate 98%
Supplier: Ambeed
|
Warning
|
A148795-100G
A148795-10G
A148795-500G
A148795-1000G
A148795-25G
103808-714EA
176
USD
103808-714
76689-270
76689-272
76689-274
77299-684
77299-686
Benzyl carbamate 98%
Benzyl carbamate
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Boiling Pt: 318.7 °C (760 mm Hg) Melting Pt: 86…89 °C Flash Pt: 182.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007965 CAS Number: 621-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...