Benzyl bis(2-chloroethyl)carbamate 95%

Supplier: Ambeed
Warning

A175871-10G A175871-1G A175871-5G A175871-100G
103807-094EA 11 USD
103807-094 103845-406 76720-658 77082-752 77082-754 77667-450
Benzyl bis(2-chloroethyl)carbamate 95%
Benzyl bis(2-chloroethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂NO₂
MW: 276.16 g/mol
Boiling Pt: 376.9 °C (760 mm Hg)
Flash Pt: 181.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06858694
CAS Number: 72791-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR