Benzyl 4-iodopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A134039-100MG A134039-250MG A134039-1G
103801-960EA 39 USD
103801-960 77010-050 77010-052 77298-684
Benzyl 4-iodopiperidine-1-carboxylate 95%
Benzyl 4-iodopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₆INO₂
MW: 345.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115043
CAS Number: 885275-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR