Benzyl-4-iodobenzoate 95%

Supplier: Ambeed
Warning

A422634-1G A422634-5G A422634-10G A422634-250MG
103827-422EA 48 USD
103827-422 103838-470 76842-094 76842-096 77033-880 77033-882
Benzyl-4-iodobenzoate 95%
Benzyl-4-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₁IO₂
Melting Pt: 64…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18072656
CAS Number: 136618-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR