Benzyl-4-hydroxypiperidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzyloxycarbonyl-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic acid Benzyl ester, 1-Cbz-4-hydroxypiperidine, 1-Carbobenzoxy-4-piperidinol, 1-Carbobenzoxy-4-hydroxypiperidine, Benzyl 4-Hydroxy-1-piperidinecarboxylate, 1-Benzyloxycarbonyl-4-piperidinol, 1-Cbz-4-piperidinol
A173100-100G
A173100-10G
A173100-250MG
A173100-500G
A173100-1G
A173100-25G
A173100-5G
103806-788EA
6
USD
103806-788
103845-082
76720-574
76720-576
76720-578
76720-580
77082-304
77082-306
77082-308
Benzyl-4-hydroxypiperidine-1-carboxylate 97%
Benzyl-4-hydroxypiperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₃ MW: 235.29 g/mol Boiling Pt: 384.9 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863722 CAS Number: 95798-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...