Benzyl-4-formylpiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl 4-formylpiperidine-1-carboxylate, 4-Formylpiperidine, N-CBZ protected

A131451-1G A131451-25G A131451-5G
103801-584EA 5 USD
103801-584 77298-486 77314-088 77314-090
Benzyl-4-formylpiperidine-1-carboxylate 95%
Benzyl-4-formylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.2897 g/mol
Boiling Pt: 384.5 °C (760 mm Hg)
Flash Pt: 186.3 °C
Storage Temperature: Freezer
MDL Number: MFCD01317806
CAS Number: 138163-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR