Benzyl-4-chlorophenyl ketone 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone
A523558-1G
A523558-25G
A523558-5G
A523558-100G
A523558-10G
103828-520EA
9
USD
103828-520
103839-634
76843-636
76843-638
76843-640
77116-700
77686-353
Benzyl-4-chlorophenyl ketone 95%
Benzyl 4-chlorophenyl ketone
Precaución: for Research Use Only
Formula:
C₁₄H₁₁ClO MW: 230.69 g/mol Boiling Pt: 365.4 °C (760 mm Hg) Melting Pt: 103…107 °C Flash Pt: 197.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016342 CAS Number: 1889-71-0 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...