3-[(Benzyloxycarbonyl)amino]propionaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-[(Benzyloxycarbonyl)amino]-1-propanal, Benzyl 3-oxopropylcarbamate, N-Phenylmethoxycarbonyl-3-aminopropanal
A323282-250MG
A323282-1G
A323282-25G
A323282-10G
103825-326EA
27
USD
103825-326
103836-262
76802-930
76839-176
76839-178
77571-914
3-[(Benzyloxycarbonyl)amino]propionaldehyde 97%
3-[(Benzyloxycarbonyl)amino]propionaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO₃ MW: 207.229 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD05664298 CAS Number: 65564-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...