Benzyl-3-oxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyloxycarbonyl-3-piperidone, Benzyl 3-oxopiperidine-1-carboxylate

A281128-25G A281128-5G A281128-100G A281128-10G A281128-500G A281128-1G
103813-540EA 5 USD
103813-540 103853-492 76837-186 76837-188 77002-168 77002-170 77002-172 77305-386
Benzyl-3-oxopiperidine-1-carboxylate 97%
Benzyl-3-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Boiling Pt: 384.1 °C (760 mm Hg)
Flash Pt: 186.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001723
CAS Number: 61995-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR