Benzyl-3-oxopiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl 3-oxopiperazine-1-carboxylate, 3-Oxopiperazine-1-carboxylic Acid Benzyl Ester, 1-Carbobenzoxy-3-oxopiperazine, 1-Cbz-3-oxopiperazine

A501172-10G A501172-250MG A501172-1G A501172-25G A501172-5G
103828-294EA 13 USD
103828-294 103839-382 76774-488 76774-490 76843-344 76843-346 76843-348
Benzyl-3-oxopiperazine-1-carboxylate 98%
Benzyl-3-oxopiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃
MW: 234.26 g/mol
Melting Pt: 118…122 °C
Flash Pt: 231.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00173924
CAS Number: 78818-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR