Benzyl-3-oxocyclobutanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-cyclobutanecarboxylic acid benzyl ester, Benzyl 3-oxocyclobutanecarboxylate

A517803-10G A517803-250MG A517803-25G A517803-1G A517803-5G
103828-480EA 25 USD
103828-480 103839-584 76818-842 76818-844 76843-588 77307-694 77307-696
Benzyl-3-oxocyclobutanecarboxylate 97%
Benzyl-3-oxocyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₃
MW: 204.23 g/mol
Boiling Pt: 324.9±35.0 °C (760 mm Hg)
Flash Pt: 143.3 °C
Storage Temperature: Ambient
MDL Number: MFCD11848894
CAS Number: 198995-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR