Benzyl-3-oxoazetidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Azetidin-3-one, N-CBZ protected, Benzyl 3-oxoazetidine-1-carboxylate
A291324-25G
A291324-5G
A291324-100G
A291324-10G
A291324-500G
A291324-1G
103813-908EA
9
USD
103813-908
103835-048
76837-640
76837-642
77032-298
77032-300
77032-302
77305-608
Benzyl-3-oxoazetidine-1-carboxylate 97%
Benzyl-3-oxoazetidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₁NO₃ MW: 205.2100 g/mol Melting Pt: 40…44 °C Flash Pt: 169.422 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09954923 CAS Number: 105258-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...