Benzyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
A179822-1G
A179822-5G
103807-476EA
10
USD
103807-476
103845-828
77301-646
77301-648
Benzyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate 97%
Benzyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₇NO₃ MW: 259.31 g/mol Boiling Pt: 415.7 °C (760 mm Hg) Flash Pt: 205.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD07776632 CAS Number: 130753-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...