Benzyl 3-hydroxy-2,2-dimethylpropanoate 97%

Supplier: Ambeed
Warning

A239634-1G A239634-10G A239634-250MG A239634-5G
103811-302EA 39 USD
103811-302 103851-100 77090-242 77098-446 77098-448 77304-168
Benzyl 3-hydroxy-2,2-dimethylpropanoate 97%
Benzyl 3-hydroxy-2,2-dimethylpropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
Boiling Pt: 117…118 °C (0.1 mm Hg)
Flash Pt: 136.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09951815
CAS Number: 17701-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR