Benzyl salicylate 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl 2-hydroxybenzoate

A171145-100G A171145-25G A171145-500G
103809-032EA 77 USD
103809-032 76690-178 77082-006 77877-123
Benzyl salicylate 98%
Benzyl salicylate

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 208
Melting Pt: 23.9
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00020034
CAS Number: 118-58-1
Merck Index: 13,01146

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR