Benzyl 2-cyano-2-(4-fluoro-5-methyl-2-nitrophenyl)acetate 97%
Supplier: Ambeed
|
Warning
|
A100503-10G
A100503-1G
A100503-5G
103795-484EA
39
USD
103795-484
103821-178
77160-160
77296-726
77296-728
Benzyl 2-cyano-2-(4-fluoro-5-methyl-2-nitrophenyl)acetate 97%
Benzyl 2-cyano-2-(4-fluoro-5-methyl-2-nitrophenyl)acetate
Precaución: for Research Use Only
Formula:
C₁₇H₁₃FN₂O₄ MW: 328.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28369447 CAS Number: 1071974-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...