Benzyl bromoacetate 98%

Supplier: Ambeed
Danger

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

A200339-100G A200339-10G A200339-500G A200339-25G A200339-5G
103847-818EA 5 USD
103847-818 77112-152 77112-154 77112-156 77302-676 77302-678
Benzyl bromoacetate 98%
Benzyl bromoacetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Boiling Pt: 287.8 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000190
CAS Number: 5437-45-6
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR