Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl 3-pyrroline-1-carboxylate, 1H-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester, benzyl 2,5-dihydropyrrole-1-carboxylate, Benzyl 2,5-dihydro-1H-pyrrole-1-carboxylate
A149172-100G
A149172-10G
A149172-1G
A149172-25G
A149172-5G
103804-010EA
13
USD
103804-010
103842-132
77028-054
77028-056
77317-616
77317-618
77317-620
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate 97%
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₃NO₂ Boiling Pt: 317.8 °C (760 mm Hg) Flash Pt: 146 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863559 CAS Number: 31970-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...