Benzyl-2,2,2-trichloroacetimidate 98%

Supplier: Ambeed
Warning

A126444-100G A126444-10G A126444-500G A126444-5G
76688-412EA 103 USD
76688-412 76688-414 76688-416 77313-024
Benzyl-2,2,2-trichloroacetimidate 98%
Benzyl-2,2,2-trichloroacetimidate

Precaución: for Research Use Only

Formula: C₉H₈Cl₃NO
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000805
CAS Number: 81927-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR