Benzyl(triphenylphosphoranylidene)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl 2-(triphenylphosphoranylidene)acetate

A115966-100G A115966-10G A115966-25G A115966-500G A115966-1G A115966-5G
103798-652EA 6 USD
103798-652 103824-516 76687-826 76687-828 76687-830 76687-832 77297-412 77311-338
Benzyl(triphenylphosphoranylidene)acetate 97%
Benzyl(triphenylphosphoranylidene)acetate

Precaución: for Research Use Only

Formula: C₂₇H₂₃O₂P
MW: 410.453 g/mol
Boiling Pt: 571.1 °C (760 mm Hg)
Melting Pt: 120…122 °C
Flash Pt: 312 °C
Storage Temperature: Ambient
MDL Number: MFCD00191787
CAS Number: 15097-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR